siRNA Details
| Gene Symbol: Gba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172692.3 | NP_766280.2 | 14, 15 | 2683 | ||
| GenBank mRNA | AK089192.1 | - | 14, 15 | 2369 | ||
| AK158174.1 | - | 12, 13 | 3054 | |||
| AK173213.1 | - | 14, 15 | 2669 | |||
| AK180481.1 | - | 1, 2 | 113 | |||
| AK188452.1 | - | 1, 2 | 130 | |||
| BC031397.1 | - | 12, 13 | 2686 | |||
| BC034105.1 | - | 11, 12 | 2434 | |||
| BC056935.1 | - | 14, 15 | 2206 | 5703478 | ||

Gba2
Genomic Map