siRNA Details
| Gene Symbol: Npr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173788.3 | NP_776149.1 | 17, 18 | 2896 | ||
| GenBank mRNA | AK158195.1 | - | 12, 13 | 1447 | ||
| BC023420.1 | - | 4, 5 | 537 | |||
| BC037458.1 | - | 12, 13 | 1352 | |||
| BC042470.1 | - | 17, 18 | 2896 | 5137261 | ||
| BC049896.1 | - | 7, 8 | 991 | |||

Npr2
Genomic Map