siRNA Details
Gene Symbol: Zfp189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289901.1 | NP_001276830.1 | 4 | 1031 | ||
NM_145547.3 | NP_663522.2 | 3 | 1202 | |||
GenBank mRNA | AK049475.1 | - | 3 | 1111 | ||
AK051265.1 | - | 3 | 1161 | |||
BC021326.1 | - | 3 | 1164 | 5065942 | ||
BC087833.1 | - | 3 | 1068 | 5708354 |
