siRNA Details
| Gene Symbol: Ikbkap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026079.3 | NP_080355.2 | 30, 31 | 3473 | ||
| GenBank mRNA | AF140786.1 | - | 30, 31 | 3401 | ||
| AF367244.1 | - | 30, 31 | 3426 | |||
| AF387811.1 | - | 30, 31 | 3440 | |||
| AK031367.1 | - | 30, 31 | 3418 | |||
| AK076152.1 | - | 3, 4 | 175 | |||
| AK147153.1 | - | 30, 31 | 3476 | |||
| AK165168.1 | - | 27, 28 | 2950 | |||
| BC035529.1 | - | 5, 6 | 514 | |||
| BC052387.1 | - | 30, 31 | 3385 | 5705589 | ||
| BC054468.1 | - | 1, 2 | 388 | |||

Ikbkap
Genomic Map