siRNA Details
| Gene Symbol: Frrs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142965.1 | NP_001136437.1 | 3 | 476 | ||
| GenBank mRNA | AK038461.1 | - | 3 | 180 | ||
| AK078331.1 | - | 2 | 139 | |||
| BC132345.1 | - | 3 | 503 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Frrs1l| Gene Symbol: Frrs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142965.1 | NP_001136437.1 | 3 | 476 | ||
| GenBank mRNA | AK038461.1 | - | 3 | 180 | ||
| AK078331.1 | - | 2 | 139 | |||
| BC132345.1 | - | 3 | 503 | |||