siRNA Details
| Gene Symbol: AI314180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172381.2 | NP_759013.2 | 17 | 1962 | ||
| GenBank mRNA | AK038775.1 | - | 18 | 1876 | ||
| AK043020.1 | - | 18 | 1879 | |||
| BC058684.1 | - | 17 | 1962 | 6834912 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI314180| Gene Symbol: AI314180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172381.2 | NP_759013.2 | 17 | 1962 | ||
| GenBank mRNA | AK038775.1 | - | 18 | 1876 | ||
| AK043020.1 | - | 18 | 1879 | |||
| BC058684.1 | - | 17 | 1962 | 6834912 | ||