siRNA Details
| Gene Symbol: Focad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081184.1 | NP_001074653.1 | 14 | 1824 | ||
| GenBank mRNA | BC057079.1 | - | 3 | 302 | ||
| BC089005.1 | - | 12 | 1491 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Focad| Gene Symbol: Focad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081184.1 | NP_001074653.1 | 14 | 1824 | ||
| GenBank mRNA | BC057079.1 | - | 3 | 302 | ||
| BC089005.1 | - | 12 | 1491 | |||