siRNA Details
| Gene Symbol: Yipf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205156.1 | NP_001192085.1 | 3 | 220 | ||
| NM_145550.3 | NP_663525.1 | 3 | 220 | |||
| GenBank mRNA | AK077502.1 | - | 3 | 197 | ||
| AK081205.1 | - | 3 | 207 | |||
| AK138108.1 | - | 3 | 215 | |||
| AK157536.1 | - | 3 | 207 | |||
| BC009080.1 | - | 3 | 152 | 2655774 | ||
| BY204210.1 | - | 3 | 221 | |||

Yipf1
Genomic Map