siRNA Details
| Gene Symbol: Slc5a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145551.4 | NP_663526.3 | 12 | 1499 | ||
| GenBank mRNA | AK029158.1 | - | 12 | 1500 | ||
| AK033425.1 | - | 11 | 1429 | |||
| AK050696.1 | - | 11 | 1379 | |||
| BC021357.1 | - | 12 | 1480 | 5012356 | ||

Slc5a9
Genomic Map