siRNA Details
| Gene Symbol: Efcab14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172698.2 | NP_766286.2 | 1 | 747 | ||
| GenBank mRNA | AK028618.1 | - | 1 | 1059 | ||
| AK028740.1 | - | 1 | 1044 | |||
| AK030011.1 | - | 1 | 1077 | |||
| AK033905.1 | - | 1 | 1041 | |||
| AK078691.1 | - | 1 | 1042 | |||
| AK079503.1 | - | 1 | 989 | |||
| AK080626.1 | - | 1 | 1043 | |||
| AK132525.1 | - | 1 | 1048 | |||
| AK163435.1 | - | 1 | 1055 | |||
| BC016074.1 | - | 1 | 389 | |||
| BC059213.1 | - | 1 | 747 | 6408754 | ||

Efcab14
Genomic Map