siRNA Details
| Gene Symbol: Efcab14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172698.2 | NP_766286.2 | 5 | 1210 | ||
| GenBank mRNA | AK028618.1 | - | 5 | 1522 | ||
| AK030011.1 | - | 5 | 1540 | |||
| AK033905.1 | - | 5 | 1504 | |||
| AK078691.1 | - | 5 | 1505 | |||
| AK079503.1 | - | 5 | 1452 | |||
| AK080626.1 | - | 5 | 1506 | |||
| AK132525.1 | - | 5 | 1511 | |||
| AK163435.1 | - | 5 | 1518 | |||
| BC016074.1 | - | 5 | 850 | |||
| BC028320.1 | - | 2 | 117 | |||
| BC059213.1 | - | 5 | 1210 | 6408754 | ||

Efcab14
Genomic Map