siRNA Details
| Gene Symbol: Atpaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181040.4 | NP_851383.2 | 8 | 985 | ||
| GenBank mRNA | AK032020.1 | - | 587 | |||
| AK217424.1 | - | 2 | 201 | |||
| BC044874.1 | - | 8 | 826 | 4487325 | ||
| CO431467.1 | - | 8 | 572 | |||

Atpaf1
Genomic Map