siRNA Details
Gene Symbol: Tmem69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177670.4 | NP_808338.2 | 3 | 276 | ||
GenBank mRNA | AK041960.1 | - | 3 | 261 | ||
BC106178.1 | - | 3 | 269 | 6512571 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177670.4 | NP_808338.2 | 3 | 276 | ||
GenBank mRNA | AK041960.1 | - | 3 | 261 | ||
BC106178.1 | - | 3 | 269 | 6512571 |