siRNA Details
| Gene Symbol: Tesk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146151.4 | NP_666263.3 | 8, 9 | 1178 | ||
| GenBank mRNA | AK031814.1 | - | 8, 9 | 1171 | ||
| AK047755.1 | - | 8, 9 | 1179 | |||
| AK053368.1 | - | 1243 | ||||
| AK134575.1 | - | 4, 5 | 284 | |||
| BC019149.1 | - | 7, 8 | 911 | 5053345 | ||
| BC029766.1 | - | 8, 9 | 1156 | 4947564 | ||

Tesk2
Genomic Map