siRNA Details
| Gene Symbol: Foxj3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290696.1 | NP_001277625.1 | 10 | 1627 | ||
| NM_172699.3 | NP_766287.1 | 11 | 1729 | |||
| GenBank mRNA | AK082840.1 | - | 11 | 1730 | ||
| AK129271.1 | - | 10 | 1600 | |||
| AK139508.1 | - | 11 | 1663 | |||
| BC058231.1 | - | 11 | 1702 | 6314981 | ||

Foxj3
Genomic Map