siRNA Details
| Gene Symbol: Zmpste24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172700.2 | NP_766288.1 | 6 | 995 | ||
| GenBank mRNA | AJ487544.1 | - | 6 | 961 | ||
| AK083566.1 | - | 6 | 995 | |||
| AK170946.1 | - | 6 | 954 | |||
| AY029194.1 | - | 6 | 751 | |||
| BC023740.1 | - | 6 | 938 | |||
| BC037610.1 | - | 1 | 45 | |||
| BC138578.1 | - | 6 | 863 | 8861598 | ||

Zmpste24
Genomic Map