siRNA Details
| Gene Symbol: Thrap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146153.3 | NP_666265.3 | 4 | 1296 | ||
| GenBank mRNA | AK034044.1 | - | 4 | 1176 | ||
| AK079601.1 | - | 4 | 1176 | |||
| BC040346.1 | - | 4 | 1227 | |||
| BC092239.1 | - | 4 | 1212 | 30659321 | ||

Thrap3
Genomic Map