siRNA Details
| Gene Symbol: 5730409E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013755.3 | NP_001013777.2 | 2 | 516 | ||
| NM_001145950.1 | NP_001139422.1 | 2 | 521 | |||
| GenBank mRNA | AK077438.1 | - | 2 | 506 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5730409E04Rik| Gene Symbol: 5730409E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013755.3 | NP_001013777.2 | 2 | 516 | ||
| NM_001145950.1 | NP_001139422.1 | 2 | 521 | |||
| GenBank mRNA | AK077438.1 | - | 2 | 506 | ||