siRNA Details
| Gene Symbol: Zfp362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081098.1 | NP_001074567.1 | 5, 6 | 863 | ||
| GenBank mRNA | BC039093.1 | - | 1, 2 | 239 | ||
| BC147222.1 | - | 4, 5 | 762 | 9056279 | ||
| BC147223.1 | - | 4, 5 | 762 | 9056281 | ||
| BC150964.1 | - | 4, 5 | 762 | 9087875 | ||

Zfp362
Genomic Map