siRNA Details
| Gene Symbol: Adgrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199696.1 | NP_001186625.1 | 2 | 667 | ||
| NM_001290714.1 | NP_001277643.1 | 2 | 875 | |||
| NM_001290715.1 | NP_001277644.1 | 1 | 437 | |||
| NM_173071.3 | NP_775094.2 | 2 | 667 | |||
| GenBank mRNA | AK147485.1 | - | 2 | 876 | ||
| AK158735.1 | - | 2 | 478 | |||
| AY168407.1 | - | 1 | 420 | |||
| BC056926.1 | - | 2 | 581 | 5705854 | ||
More Information
Set Membership: |
|

Adgrb2
Genomic Map