siRNA Details
| Gene Symbol: Themis2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033308.2 | NP_001028480.1 | 3 | 470 | ||
| GenBank mRNA | AK137465.1 | - | 3 | 475 | ||
| AK149775.1 | - | 3 | 475 | |||
| AK150471.1 | - | 3 | 471 | |||
| AK153536.1 | - | 3 | 471 | |||
| AK155536.1 | - | 3 | 490 | |||
| AK171009.1 | - | 3 | 475 | |||
| AK171858.1 | - | 3 | 487 | |||
| BC013712.1 | - | 3 | 464 | |||
| BC151122.1 | - | 3 | 519 | 9088035 | ||
| BC151126.1 | - | 3 | 519 | 9088039 | ||

Themis2
Genomic Map