siRNA Details
| Gene Symbol: Fam76a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163792.1 | NP_001157264.1 | 2, 3 | 249 | ||
| NM_145553.2 | NP_663528.1 | 2, 3 | 249 | |||
| GenBank mRNA | AK038831.1 | - | 2, 3 | 257 | ||
| AK154575.1 | - | 2, 3 | 250 | |||
| BC008163.1 | - | 2, 3 | 209 | 3709256 | ||

Fam76a
Genomic Map