siRNA Details
| Gene Symbol: Pigv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145955.1 | NP_001139427.1 | 4 | 887 | ||
| NM_001145956.1 | NP_001139428.1 | 3 | 611 | |||
| NM_178698.5 | NP_848813.3 | 4 | 1733 | |||
| GenBank mRNA | AK046527.1 | - | 4 | 1694 | ||
| AK047322.1 | - | 4 | 1531 | |||
| AK052446.1 | - | 4 | 1707 | |||
| AK080590.1 | - | 3 | 612 | |||
| AK080858.1 | - | 4 | 1687 | |||
| AK148285.1 | - | 3 | 600 | |||
| BC055060.1 | - | 4 | 1683 | 6528864 | ||

Pigv
Genomic Map