siRNA Details
| Gene Symbol: Pigv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178698.5 | NP_848813.3 | 3 | 581 | ||
| GenBank mRNA | AK046527.1 | - | 3 | 542 | ||
| AK047322.1 | - | 3 | 379 | |||
| AK052446.1 | - | 3 | 555 | |||
| AK080858.1 | - | 3 | 535 | |||
| BC055060.1 | - | 3 | 531 | 6528864 | ||

Pigv
Genomic Map