siRNA Details
| Gene Symbol: Slc30a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039677.2 | NP_001034766.1 | 6 | 1039 | ||
| GenBank mRNA | AK031425.1 | - | 6 | 835 | ||
| BC013478.1 | - | 5 | 1059 | |||
| BC113751.1 | - | 6 | 1039 | 40057211 | ||
| BC147704.1 | - | 6 | 870 | 9087463 | ||
| BC147707.1 | - | 6 | 870 | 9087466 | ||

Slc30a2
Genomic Map