siRNA Details
| Gene Symbol: Asap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008232.2 | NP_001008233.1 | 5 | 606 | ||
| GenBank mRNA | AK132294.1 | - | 5 | 575 | ||
| BC085250.1 | - | 5 | 559 | 30730459 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asap3| Gene Symbol: Asap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008232.2 | NP_001008233.1 | 5 | 606 | ||
| GenBank mRNA | AK132294.1 | - | 5 | 575 | ||
| BC085250.1 | - | 5 | 559 | 30730459 | ||