siRNA Details
| Gene Symbol: Zbtb40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198248.1 | NP_937891.1 | 17 | 3435 | ||
| GenBank mRNA | AK048974.1 | - | 2 | 91 | ||
| AK220256.1 | - | 18 | 3320 | |||
| BC059177.1 | - | 17 | 3435 | 6410689 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb40| Gene Symbol: Zbtb40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198248.1 | NP_937891.1 | 17 | 3435 | ||
| GenBank mRNA | AK048974.1 | - | 2 | 91 | ||
| AK220256.1 | - | 18 | 3320 | |||
| BC059177.1 | - | 17 | 3435 | 6410689 | ||