siRNA Details
| Gene Symbol: Emc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039200.2 | NP_001034289.1 | 21, 22 | 2715 | ||
| NM_146157.4 | NP_666269.2 | 22, 23 | 2724 | |||
| GenBank mRNA | AF525925.1 | - | 12, 13 | 1563 | ||
| AK049061.1 | - | 22, 23 | 2715 | |||
| AK077496.1 | - | 22, 23 | 2715 | |||
| AK129052.1 | - | 22, 23 | 2677 | |||
| AK161677.1 | - | 21, 22 | 2680 | |||
| BC056952.1 | - | 17, 18 | 2444 | |||
| BC129804.1 | - | 22, 23 | 2711 | 40087024 | ||
| BC139366.1 | - | 22, 23 | 2710 | 8862388 | ||

Emc1
Genomic Map