siRNA Details
| Gene Symbol: Cep104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177673.2 | NP_808341.1 | 17 | 2305 | ||
| GenBank mRNA | AK129163.1 | - | 16 | 2171 | ||
| AK143802.1 | - | 17 | 2722 | |||
| AK149826.1 | - | 17 | 2656 | |||
| BC023302.1 | - | 2 | 68 | |||
| BC046331.1 | - | 17 | 2305 | 6485859 | ||
| BC060106.1 | - | 10 | 1497 | |||

Cep104
Genomic Map