siRNA Details
| Gene Symbol: Hadhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289798.1 | NP_001276727.1 | 10 | 1008 | ||
| NM_001289799.1 | NP_001276728.1 | 10 | 1004 | |||
| NM_145558.2 | NP_663533.1 | 10 | 961 | |||
| GenBank mRNA | AK033462.1 | - | 10 | 943 | ||
| AK076814.1 | - | 10 | 924 | |||
| AK083164.1 | - | 10 | 969 | |||
| AK083767.1 | - | 10 | 924 | |||
| AK150889.1 | - | 10 | 923 | |||
| AK168594.1 | - | 10 | 925 | |||
| AK169637.1 | - | 10 | 937 | |||
| BC005585.1 | - | 9 | 892 | 3158015 | ||

Hadhb
Genomic Map