siRNA Details
| Gene Symbol: Hadhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289798.1 | NP_001276727.1 | 9 | 931 | ||
| NM_001289799.1 | NP_001276728.1 | 9 | 927 | |||
| NM_145558.2 | NP_663533.1 | 9 | 884 | |||
| GenBank mRNA | AK033462.1 | - | 9 | 866 | ||
| AK076814.1 | - | 9 | 847 | |||
| AK083164.1 | - | 9 | 892 | |||
| AK083767.1 | - | 9 | 847 | |||
| AK150889.1 | - | 9 | 846 | |||
| AK168594.1 | - | 9 | 848 | |||
| AK169637.1 | - | 9 | 860 | |||
| BC005585.1 | - | 8 | 815 | 3158015 | ||

Hadhb
Genomic Map