siRNA Details
| Gene Symbol: Fam193a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243123.1 | NP_001230052.1 | 14 | 2693 | ||
| GenBank mRNA | AK144988.1 | - | 13 | 2210 | ||
| BC037112.1 | - | 11 | 1830 | |||
| BC099925.1 | - | 11 | 1730 | 30653351 | ||

Fam193a
Genomic Map