siRNA Details
| Gene Symbol: Dok7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172708.3 | NP_766296.1 | 8 | 1548 | ||
| GenBank mRNA | AK044445.1 | - | 8 | 1549 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dok7| Gene Symbol: Dok7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172708.3 | NP_766296.1 | 8 | 1548 | ||
| GenBank mRNA | AK044445.1 | - | 8 | 1549 | ||