siRNA Details
| Gene Symbol: Sh3tc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194344.2 | NP_919325.2 | 6 | 983 | ||
| GenBank mRNA | AK028482.1 | - | 2 | 1079 | ||
| AK150064.1 | - | 6 | 761 | |||
| AK155330.1 | - | 6 | 744 | |||
| AK159089.1 | - | 6 | 762 | |||
| AK167283.1 | - | 6 | 684 | |||
| AK170383.1 | - | 6 | 816 | |||
| AK170396.1 | - | 6 | 763 | |||
| AK171209.1 | - | 6 | 740 | |||
| AK171242.1 | - | 6 | 740 | |||
| BC024909.1 | - | 4 | 433 | |||
| BC056461.1 | - | 4 | 327 | |||

Sh3tc1
Genomic Map