siRNA Details
| Gene Symbol: Sh3tc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194344.2 | NP_919325.2 | 12 | 2919 | ||
| GenBank mRNA | AK028482.1 | - | 8 | 3015 | ||
| AK131143.1 | - | 6 | 1784 | |||
| AK150064.1 | - | 12 | 2697 | |||
| AK155330.1 | - | 12 | 2685 | |||
| AK159089.1 | - | 12 | 2698 | |||
| AK167283.1 | - | 12 | 2620 | |||
| AK170383.1 | - | 12 | 2752 | |||
| BC024909.1 | - | 10 | 2369 | |||
| BC026441.1 | - | 1 | 335 | |||
| BC056461.1 | - | 10 | 2263 | |||

Sh3tc1
Genomic Map