siRNA Details
| Gene Symbol: Cc2d2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172274.2 | NP_758478.1 | 9 | 1000 | ||
| GenBank mRNA | AK077658.1 | - | 6 | 475 | ||
| AK145128.1 | - | 6 | 538 | |||
| BC032290.1 | - | 9 | 944 | 5369681 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cc2d2a| Gene Symbol: Cc2d2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172274.2 | NP_758478.1 | 9 | 1000 | ||
| GenBank mRNA | AK077658.1 | - | 6 | 475 | ||
| AK145128.1 | - | 6 | 538 | |||
| BC032290.1 | - | 9 | 944 | 5369681 | ||