siRNA Details
| Gene Symbol: Sel1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172710.3 | NP_766298.2 | 15 | 2512 | ||
| GenBank mRNA | AK030487.1 | - | 15 | 2377 | ||
| AK122362.1 | - | 15 | 2512 | |||
| BC026655.1 | - | 3 | 235 | |||
| BC094607.1 | - | 14 | 2057 | 6837048 | ||
| BC117915.1 | - | 14 | 2013 | 40097358 | ||
| BC117916.1 | - | 14 | 2015 | 40097359 | ||

Sel1l3
Genomic Map