siRNA Details
| Gene Symbol: Sel1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172710.3 | NP_766298.2 | 4 | 1076 | ||
| GenBank mRNA | AK030487.1 | - | 4 | 941 | ||
| AK037220.1 | - | 4 | 940 | |||
| AK122362.1 | - | 4 | 1076 | |||
| BC094607.1 | - | 3 | 621 | 6837048 | ||
| BC117915.1 | - | 3 | 577 | 40097358 | ||
| BC117916.1 | - | 3 | 579 | 40097359 | ||

Sel1l3
Genomic Map