siRNA Details
| Gene Symbol: Chrna9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081104.1 | NP_001074573.1 | 4 | 908 | ||
| GenBank mRNA | AY095483.1 | - | 1 | 78 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrna9| Gene Symbol: Chrna9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081104.1 | NP_001074573.1 | 4 | 908 | ||
| GenBank mRNA | AY095483.1 | - | 1 | 78 | ||