siRNA Details
| Gene Symbol: Atp10d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153389.3 | NP_700438.3 | 9 | 1449 | ||
| GenBank mRNA | AJ441079.1 | - | 9 | 1449 | ||
| AK036646.1 | - | 8 | 1114 | |||
| BC141328.1 | - | 9 | 1300 | 9055940 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp10d| Gene Symbol: Atp10d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153389.3 | NP_700438.3 | 9 | 1449 | ||
| GenBank mRNA | AJ441079.1 | - | 9 | 1449 | ||
| AK036646.1 | - | 8 | 1114 | |||
| BC141328.1 | - | 9 | 1300 | 9055940 | ||