siRNA Details
Gene Symbol: Tmem150c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182841.1 | NP_878261.1 | 8 | 1048 | ||
GenBank mRNA | AK044570.1 | - | 8 | 809 | ||
BC028943.1 | - | 3 | 369 | |||
BC055321.1 | - | 8 | 1048 | 6419443 | ||
BC062109.1 | - | 8 | 826 |
