siRNA Details
| Gene Symbol: Agpat9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172715.3 | NP_766303.1 | 8 | 1127 | ||
| GenBank mRNA | AK030171.1 | - | 9 | 1312 | ||
| AK138410.1 | - | 7 | 1287 | |||
| AK160261.1 | - | 8 | 1072 | |||
| BC096769.1 | - | 8 | 1060 | 30101305 | ||
| BC138228.1 | - | 7 | 880 | 8861247 | ||
| BC145669.1 | - | 7 | 880 | 40130972 | ||
| CN719213.1 | - | 4 | 338 | |||

Agpat9
Genomic Map