siRNA Details
Gene Symbol: Lrrc8d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122768.1 | NP_001116240.1 | 3 | 951 | ||
NM_178701.3 | NP_848816.3 | 3 | 941 | |||
GenBank mRNA | AK046249.1 | - | 3 | 950 | ||
AK050400.1 | - | 3 | 911 | |||
AK080722.1 | - | 3 | 885 | |||
AK137459.1 | - | 3 | 952 | |||
AK167985.1 | - | 2 | 659 | |||
BC037717.1 | - | 3 | 922 | 4239389 |
