siRNA Details
| Gene Symbol: Lrrc8d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122768.1 | NP_001116240.1 | 3 | 2963 | ||
| NM_178701.3 | NP_848816.3 | 3 | 2953 | |||
| GenBank mRNA | AK046249.1 | - | 3 | 2962 | ||
| AK050400.1 | - | 3 | 2923 | |||
| AK080722.1 | - | 3 | 2897 | |||
| AK137459.1 | - | 3 | 2964 | |||
| AK167985.1 | - | 2 | 2671 | |||
| AK201741.1 | - | 1 | 324 | |||
| AK212520.1 | - | 1 | 323 | |||
| BC014756.1 | - | 1 | 1987 | |||
| BC037717.1 | - | 3 | 2934 | 4239389 | ||

Lrrc8d
Genomic Map