siRNA Details
| Gene Symbol: Oasl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145209.3 | NP_660210.1 | 3 | 404 | ||
| GenBank mRNA | AB067533.1 | - | 2 | 329 | ||
| AF426289.1 | - | 2 | 335 | |||
| AK078690.1 | - | 3 | 405 | |||
| AK089111.1 | - | 2 | 374 | |||
| AK149824.1 | - | 2 | 328 | |||
| AK150863.1 | - | 2 | 458 | |||
| AK165578.1 | - | 2 | 441 | |||
| AY057107.1 | - | 2 | 317 | |||
| AY089728.1 | - | 2 | 329 | |||
| BC032152.1 | - | 2 | 341 | 5068699 | ||

Oasl1
Genomic Map