siRNA Details
Gene Symbol: Naa25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172722.3 | NP_766310.2 | 10 | 1001 | ||
GenBank mRNA | AK049325.1 | - | 10 | 1002 | ||
AK052060.1 | - | 10 | 989 | |||
AK149758.1 | - | 10 | 1002 | |||
AK165175.1 | - | 10 | 992 | |||
AK167405.1 | - | 10 | 1019 | |||
AK167491.1 | - | 10 | 1019 | |||
BC028868.1 | - | 8 | 781 | |||
BC037595.1 | - | 10 | 982 | |||
BC055480.1 | - | 9 | 903 | |||
BC082314.1 | - | 10 | 987 | |||
BC106182.1 | - | 10 | 976 | |||
BC132172.1 | - | 10 | 999 | 40130449 | ||
BC138093.1 | - | 10 | 999 | 8861112 |
