siRNA Details
Gene Symbol: Naa25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172722.3 | NP_766310.2 | 16 | 1804 | ||
GenBank mRNA | AK041906.1 | - | 2 | 121 | ||
AK049325.1 | - | 16 | 1805 | |||
AK149758.1 | - | 16 | 1805 | |||
AK165175.1 | - | 16 | 1795 | |||
AK167405.1 | - | 16 | 1822 | |||
AK167491.1 | - | 16 | 1822 | |||
BC028868.1 | - | 14 | 1584 | |||
BC037595.1 | - | 16 | 1785 | |||
BC055480.1 | - | 15 | 1706 | |||
BC082314.1 | - | 16 | 1790 | |||
BC106182.1 | - | 16 | 1779 | |||
BC132172.1 | - | 16 | 1802 | 40130449 | ||
BC138093.1 | - | 16 | 1802 | 8861112 |
Gene Symbol: Erp29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077472.1 | - | 1963 |
