siRNA Details
| Gene Symbol: Rimbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081388.1 | NP_001074857.1 | 11 | 1583 | ||
| GenBank mRNA | AK038814.1 | - | 11 | 1277 | ||
| AK044685.1 | - | 10 | 1683 | |||
| AK122245.2 | - | 2 | 396 | |||
| AK147417.1 | - | 7 | 987 | |||
| BC151100.1 | - | 7 | 1174 | 9088013 | ||
| BC151101.1 | - | 7 | 1174 | 9088014 | ||

Rimbp2
Genomic Map