siRNA Details
| Gene Symbol: Tmem184a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161548.1 | NP_001155020.1 | 9 | 1262 | ||
| NM_144914.3 | NP_659163.3 | 8 | 1082 | |||
| GenBank mRNA | AK148454.1 | - | 1054 | |||
| BC019731.1 | - | 4 | 471 | 3993397 | ||
| BC026659.1 | - | 9 | 1221 | 4988857 | ||

Tmem184a
Genomic Map