siRNA Details
| Gene Symbol: Tmem184a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161548.1 | NP_001155020.1 | 9 | 1486 | ||
| NM_144914.3 | NP_659163.3 | 8 | 1306 | |||
| GenBank mRNA | AK050489.1 | - | 9 | 1456 | ||
| AK148454.1 | - | 1278 | ||||
| BC019731.1 | - | 4 | 695 | 3993397 | ||
| BC026659.1 | - | 9 | 1445 | 4988857 | ||

Tmem184a
Genomic Map